5-Bromo-4-chloro-2-methoxypyridine 98%

Supplier: Ambeed
Warning

A248239-100MG A248239-250MG A248239-1G
103811-818EA 26 USD
103811-818 103851-652 77031-438 77031-440 77090-992
5-Bromo-4-chloro-2-methoxypyridine 98%
5-Bromo-4-chloro-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅BrClNO
MW: 222.467 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17014994
CAS Number: 851607-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR