(5-Bromo-3-pyridinyl)methanol 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-5-(hydroxymethyl)pyridine, 5-Bromo-3-pyridinemethanol

A107079-100G A107079-10G A107079-1G A107079-5G
103797-120EA 8 USD
103797-120 103822-878 77160-534 77160-536 77309-750 77309-752
(5-Bromo-3-pyridinyl)methanol 98%
(5-Bromo-3-pyridinyl)methanol

Precaución: for Research Use Only

Formula: C₆H₆BrNO
MW: 188.0200 g/mol
Boiling Pt: 291.5 °C (760 mm Hg)
Melting Pt: 33…37 °C
Flash Pt: 130.1 °C
Storage Temperature: Ambient
MDL Number: MFCD03265757
CAS Number: 37669-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR