5-Bromo-3-nitropyridine-2-carbonitrile 98%

Supplier: Ambeed
Danger

A221014-25G A221014-5G A221014-1G A221014-100G
103818-882EA 9 USD
103818-882 103849-762 77088-352 77088-354 77303-568 77748-355
5-Bromo-3-nitropyridine-2-carbonitrile 98%
5-Bromo-3-nitropyridine-2-carbonitrile

Precaución: for Research Use Only

Formula: C₆H₂BrN₃O₂
Boiling Pt: 348.9 °C (760 mm Hg)
Melting Pt: 101…106 °C
Storage Temperature: Ambient
MDL Number: MFCD06657551
CAS Number: 573675-25-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR