5-Bromo-3-methyl-1H-indazole 98%

Supplier: Ambeed
Warning

A101043-250MG A101043-25G A101043-1G A101043-10G
103795-776EA 9 USD
103795-776 103821-466 77160-188 77160-190 77296-964 77570-080
5-Bromo-3-methyl-1H-indazole 98%
5-Bromo-3-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇BrN₂
MW: 211.0600 g/mol
Boiling Pt: 341.4 °C (760 mm Hg)
Flash Pt: 160.284 °C
Storage Temperature: Ambient
MDL Number: MFCD05664003
CAS Number: 552331-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR