3-Methyl-5-bromo-1,2,4-thiadiazole 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A117462-250MG A117462-5G A117462-100MG
103798-892EA 54 USD
103798-892 103824-780 76822-200 77311-562 77683-057
3-Methyl-5-bromo-1,2,4-thiadiazole 98%, Ambeed.Inc
3-Methyl-5-bromo-1,2,4-thiadiazole

Precaución: For research use only.

Formula: C₃H₃BrN₂S
Melting Pt: 28…31 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11847500
CAS Number: 54681-68-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR