5-Bromo-3-methoxypicolinic acid 95%
Supplier: Ambeed
|
Warning
|
A199667-250MG
A199667-1G
A199667-5G
103817-124EA
60
USD
103817-124
103847-750
76999-274
77085-798
77085-800
5-Bromo-3-methoxypicolinic acid 95%
5-Bromo-3-methoxypicolinic acid
Precaución: for Research Use Only
Formula:
C₇H₆BrNO₃ MW: 232.0300 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11857696 CAS Number: 1142191-66-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...