5-Bromo-3-methoxy-2-nitropyridine 98%
Supplier: Ambeed
|
Warning
|
A167989-250MG
A167989-1G
103806-238EA
30
USD
103806-238
103844-496
76769-016
77081-534
5-Bromo-3-methoxy-2-nitropyridine 98%
5-Bromo-3-methoxy-2-nitropyridine
Precaución: for Research Use Only
Formula:
C₆H₅BrN₂O₃ MW: 233.02 g/mol Boiling Pt: 334.6 °C (760 mm Hg) Melting Pt: 112 °C Flash Pt: 156.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00834963 CAS Number: 152684-26-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...