5-Bromo-3-isopropyl-1H-indazole 95%

Supplier: Ambeed
Warning

A140653-100MG A140653-250MG A140653-1G A140653-5G
103802-890EA 329 USD
103802-890 77162-298 77162-300 77299-154 77299-156
5-Bromo-3-isopropyl-1H-indazole 95%
5-Bromo-3-isopropyl-1H-indazole

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrN₂
Storage Temperature: Ambient
MDL Number: MFCD22034709
CAS Number: 1276075-40-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR