5-Bromo-3-iodo-2-hydroxypyridine 95%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-3-iodopyridin-2(1H)-one, 5-Bromo-2-hydroxy-3-iodopyridine

A103707-10G A103707-250MG A103707-25G A103707-5G A103707-1G
103796-600EA 7 USD
103796-600 103822-310 77008-508 77008-510 77008-512 77283-146 77309-178
5-Bromo-3-iodo-2-hydroxypyridine 95%
5-Bromo-3-iodo-2-hydroxypyridine

Precaución: for Research Use Only

Formula: C₅H₃BrINO
MW: 299.89 g/mol
Boiling Pt: 316.3 °C (760 mm Hg)
Melting Pt: 226…227 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08688586
CAS Number: 381233-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR