5-Bromo-3-iodo-4,7-diazaindole 95%

Supplier: Ambeed
Warning

A157699-100MG A157699-250MG A157699-1G A157699-5G
103805-036EA 40 USD
103805-036 103843-238 77259-490 77259-492 77300-282 77300-284
5-Bromo-3-iodo-4,7-diazaindole 95%
5-Bromo-3-iodo-4,7-diazaindole

Precaución: for Research Use Only

Formula: C₆H₃BrIN₃
MW: 323.9200 g/mol
Boiling Pt: 311 °C (760 mm Hg)
Flash Pt: 142 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09878696
CAS Number: 875781-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR