5-Bromo-3-iodo-1H-pyrrolo[2,3-b]pyridine 95%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-3-iodo-7-azaindole

A101482-10G A101482-25G A101482-1G A101482-5G A101482-100G
103795-998EA 9 USD
103795-998 103821-682 77160-200 77160-202 77297-124 77297-126 77592-070
5-Bromo-3-iodo-1H-pyrrolo[2,3-b]pyridine 95%
5-Bromo-3-iodo-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₄BrIN₂
MW: 322.93 g/mol
Melting Pt: 234…239 °C
Flash Pt: 191.1 °C
Storage Temperature: Ambient
MDL Number: MFCD07779519
CAS Number: 757978-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR