5-Bromo-3-fluoropyridin-2-amine 98%

Supplier: Ambeed
Warning

A359879-100G A359879-10G A359879-250MG A359879-1G A359879-25G A359879-5G
103826-240EA 6 USD
103826-240 103837-248 76788-546 76788-548 76788-550 76840-440 76840-442 76840-444
5-Bromo-3-fluoropyridin-2-amine 98%
5-Bromo-3-fluoropyridin-2-amine

Precaución: for Research Use Only

Formula: C₅H₄BrFN₂
Flash Pt: 73.7 °C
Storage Temperature: Freezer
MDL Number: MFCD11848483
CAS Number: 748812-37-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR