5-Bromo-3-fluoro-2-hydroxypyridine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-3-fluoropyridin-2(1H)-one

A114028-10G A114028-1G A114028-5G
103798-332EA 15 USD
103798-332 103824-174 76995-416 77297-286 77297-288
5-Bromo-3-fluoro-2-hydroxypyridine 98%
5-Bromo-3-fluoro-2-hydroxypyridine

Precaución: for Research Use Only

Formula: C₅H₃BrFNO
MW: 191.986 g/mol
Boiling Pt: 274.1 °C (760 mm Hg)
Flash Pt: 119.6 °C
Storage Temperature: Ambient
MDL Number: MFCD07375017
CAS Number: 156772-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR