5-Bromo-3-fluoro-2-nitropyridine 97%

Supplier: Ambeed
Warning

A163938-100MG A163938-250MG
103805-792EA 41 USD
103805-792 103844-058 77096-234 77096-236
5-Bromo-3-fluoro-2-nitropyridine 97%
5-Bromo-3-fluoro-2-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrFN₂O₂
MW: 220.98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28129784
CAS Number: 1532517-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR