5-Bromo-3-fluoro-2-methoxypyridine 98%
Supplier: Ambeed
|
Warning
|
A136053-100G
A136053-10G
A136053-250MG
A136053-1G
A136053-25G
A136053-5G
103802-266EA
9
USD
103802-266
77010-148
77010-150
77010-152
77315-150
77315-152
77315-154
5-Bromo-3-fluoro-2-methoxypyridine 98%
5-Bromo-3-fluoro-2-methoxypyridine
Precaución: for Research Use Only
Formula:
C₆H₅BrFNO Boiling Pt: 189.6 °C (760 mm Hg) Melting Pt: 40…43 °C Flash Pt: 68.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD04112553 CAS Number: 124432-70-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...