5-Bromo-3-cyanoindole 97%

Supplier: Ambeed
Danger

Synonyms: 5-Bromo-1H-indole-3-carbonitrile

A210636-1G A210636-5G A210636-100MG A210636-10G A210636-250MG A210636-25G
103818-028EA 10 USD
103818-028 103848-722 77087-104 77087-106 77139-590 77139-592 77139-594 77139-596
5-Bromo-3-cyanoindole 97%
5-Bromo-3-cyanoindole

Precaución: for Research Use Only

Formula: C₉H₅BrN₂
MW: 221.0500 g/mol
Boiling Pt: 405.9 °C (760 mm Hg)
Flash Pt: 199.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04127133
CAS Number: 90271-86-6
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR