5-Bromo-3-chloropyridin-2(1H)-one 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-3-chloro-2-hydroxypyridine

A168961-100G A168961-10G A168961-250MG A168961-25G A168961-1G A168961-5G
103806-336EA 8 USD
103806-336 103844-592 76813-140 76813-142 76813-144 76813-146 77081-660 77081-662
5-Bromo-3-chloropyridin-2(1H)-one 98%
5-Bromo-3-chloropyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₅H₃BrClNO
MW: 208.44 g/mol
Boiling Pt: 303.8 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD05662389
CAS Number: 58236-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR