5-Bromo-3-chloropyrazin-2-amine 95%

Supplier: Ambeed
Warning

A286635-100G A286635-10G A286635-1G A286635-25G
103813-724EA 13 USD
103813-724 103834-860 76787-468 76787-470 76837-402 77305-500
5-Bromo-3-chloropyrazin-2-amine 95%
5-Bromo-3-chloropyrazin-2-amine

Precaución: for Research Use Only

Formula: C₄H₃BrClN₃
MW: 208.4500 g/mol
Boiling Pt: 271.3 °C (760 mm Hg)
Melting Pt: 128 °C
Flash Pt: 118 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11109828
CAS Number: 21943-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR