5-Bromo-3-chloropyrazin-2(1H)-one 97%

Supplier: Ambeed
Warning

A109822-250MG A109822-25G A109822-5G A109822-1G
103797-566EA 35 USD
103797-566 103823-372 77245-634 77245-636 77283-886 77310-180
5-Bromo-3-chloropyrazin-2(1H)-one 97%
5-Bromo-3-chloropyrazin-2(1H)-one

Precaución: for Research Use Only

Formula: C₄H₂BrClN₂O
Storage Temperature: Refrigerator
MDL Number: MFCD28369353
CAS Number: 21943-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR