5-Bromo-3-chloropyridine-2-carbonitrile 98%

Supplier: Ambeed
Warning

A195956-10G A195956-250MG A195956-1G A195956-25G A195956-5G
103816-796EA 15 USD
103816-796 103847-410 77029-982 77029-984 77085-360 77085-362 77085-364
5-Bromo-3-chloropyridine-2-carbonitrile 98%
5-Bromo-3-chloropyridine-2-carbonitrile

Precaución: for Research Use Only

Formula: C₆H₂BrClN₂
Storage Temperature: Ambient
MDL Number: MFCD12827550
CAS Number: 945557-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR