5-Bromo-3-chloropyridine-2-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Bromo-3-chloropicolinic acid
A333260-1G
A333260-25G
A333260-100G
A333260-10G
A333260-250MG
103825-628EA
10
USD
103825-628
103836-592
76839-600
76839-602
77032-910
77032-912
77032-914
5-Bromo-3-chloropyridine-2-carboxylic acid 98%
5-Bromo-3-chloropyridine-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₆H₃BrClNO₂ MW: 236.46 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD12827552 CAS Number: 1189513-51-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...