5-Bromo-3-chloropicolinaldehyde 95%

Supplier: Ambeed
Warning

A193811-250MG A193811-1G A193811-5G
103816-584EA 191 USD
103816-584 103847-196 77192-072 77302-362 77302-364
5-Bromo-3-chloropicolinaldehyde 95%
5-Bromo-3-chloropicolinaldehyde

Precaución: Inert atmosphere, 2…8 °C.

Formula: C₆H₃BrClNO
Storage Temperature: Refrigerator
MDL Number: MFCD08277307
CAS Number: 885168-04-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR