5-Bromo-3-chloro-2-fluoropyridine 97%

Supplier: Ambeed
Warning

A155474-1G A155474-25G A155474-5G
103804-764EA 5 USD
103804-764 103842-962 77318-684 77318-686 77318-688
5-Bromo-3-chloro-2-fluoropyridine 97%
5-Bromo-3-chloro-2-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrClFN
MW: 210.43 g/mol
Flash Pt: 78.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09834365
CAS Number: 38185-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR