5-Bromo-3-chloro-2-fluorophenol 95%

Supplier: Ambeed
Warning

A107901-250MG A107901-1G
103797-262EA 26 USD
103797-262 103823-032 76869-526 77309-880
5-Bromo-3-chloro-2-fluorophenol 95%
5-Bromo-3-chloro-2-fluorophenol

Precaución: for Research Use Only

Formula: C₆H₃BrClFO
MW: 225.4428 g/mol
Boiling Pt: 240.8±35.0 °C (760 mm Hg)
Flash Pt: 99.4 °C
Storage Temperature: Ambient
MDL Number: MFCD18914516
CAS Number: 1305322-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR