5-Bromo-3-chloro-1H-pyrrolo[2,3-b]pyridine 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-3-chloro-7-azaindole

A895245-25G A895245-10G A895245-250MG A895245-1G A895245-5G
103841-334EA 38 USD
103841-334 76845-596 76880-768 76880-770 77308-492 77308-494
5-Bromo-3-chloro-1H-pyrrolo[2,3-b]pyridine 97%
5-Bromo-3-chloro-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₄BrClN₂
MW: 231.4771 g/mol
Melting Pt: 216…220 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09817420
CAS Number: 951626-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR