5-Bromo-3-chloro-1H-indazole 97%

Supplier: Ambeed
Warning

A733512-1G A733512-10G
103840-794EA 89 USD
103840-794 76845-016 77170-944
5-Bromo-3-chloro-1H-indazole 97%
5-Bromo-3-chloro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄BrClN₂
MW: 231.4800 g/mol
Boiling Pt: 368.5 °C (760 mm Hg)
Flash Pt: 176.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06738284
CAS Number: 36760-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR