5-Bromo-3,4-dihydroquinolin-2(1H)-one 97%

Supplier: Ambeed
Warning

A122666-100MG A122666-250MG A122666-1G
103800-252EA 72 USD
103800-252 76766-596 76766-598 77312-382
5-Bromo-3,4-dihydroquinolin-2(1H)-one 97%
5-Bromo-3,4-dihydroquinolin-2(1H)-one

Precaución: for Research Use Only

Formula: C₉H₈BrNO
Storage Temperature: Ambient
MDL Number: MFCD11036287
CAS Number: 880094-83-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR