5-Bromo-3,4-dihydroisoquinolin-1(2H)-one 97%
Supplier: Ambeed
|
Warning
|
A131182-100MG
A131182-250MG
103801-536EA
55
USD
103801-536
76972-062
76972-064
5-Bromo-3,4-dihydroisoquinolin-1(2H)-one 97%
5-Bromo-3,4-dihydroisoquinolin-1(2H)-one
Precaución: for Research Use Only
Formula:
C₉H₈BrNO Melting Pt: 180…183 °C Storage Temperature: Ambient |
MDL Number:
MFCD11847788 CAS Number: 1109230-25-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...