5-Bromo-3,4-dihydro-2H-benzo[b][1,4]oxazine 95%

Supplier: Ambeed
Warning

A213768-100MG A213768-250MG
103818-338EA 323 USD
103818-338 76647-532 76647-534
5-Bromo-3,4-dihydro-2H-benzo[b][1,4]oxazine 95%
5-Bromo-3,4-dihydro-2H-benzo[b][1,4]oxazine

Precaución: for Research Use Only

Formula: C₈H₈BrNO
MW: 214.05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31698815
CAS Number: 1402672-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR