5-Bromo-2-cyano-3-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Warning

A148253-250MG A148253-1G A148253-5G
103803-900EA 37 USD
103803-900 103842-016 77027-984 77299-646 77299-648
5-Bromo-2-cyano-3-(trifluoromethyl)pyridine 98%
5-Bromo-2-cyano-3-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₂BrF₃N₂
Boiling Pt: 257 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD13185358
CAS Number: 1214377-57-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR