5-Bromo-3-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine ≥95%
Supplier: Ambeed
|
Warning
|
A102966-250MG
A102966-5G
103796-454EA
220
USD
103796-454
77610-778
77882-878
5-Bromo-3-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine ≥95%
5-Bromo-3-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine
Precaución: Sealed in dry, at room temperature.
Formula:
C₈H₄BrF₃N₂ Storage Temperature: Ambient |
MDL Number:
MFCD12024503 CAS Number: 1150618-36-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...