5-Bromo-3-(difluoromethoxy)pyridin-2-amine 98%

Supplier: Ambeed
Warning

A232237-10G A232237-250MG A232237-1G A232237-5G
103810-826EA 70 USD
103810-826 103850-578 77031-080 77031-082 77303-936 77303-938
5-Bromo-3-(difluoromethoxy)pyridin-2-amine 98%
5-Bromo-3-(difluoromethoxy)pyridin-2-amine

Precaución: for Research Use Only

Formula: C₆H₅BrF₂N₂O
MW: 239.02 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12405789
CAS Number: 947249-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR