5-Bromo-2-phenyl-1H-indole 97%

Supplier: Ambeed
Warning

A214290-1G A214290-5G A214290-250MG
103818-388EA 92 USD
103818-388 103849-094 77087-608 77087-610 77237-498
5-Bromo-2-phenyl-1H-indole 97%
5-Bromo-2-phenyl-1H-indole

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrN
MW: 272.14 g/mol
Melting Pt: 195…196 °C
Storage Temperature: Ambient
MDL Number: MFCD04151729
CAS Number: 83515-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR