5-Bromo-2-oxo-2,3-dihydropyridine-3-carbonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-hydroxynicotinonitrile

A213364-10G A213364-250MG A213364-1G A213364-25G A213364-5G
103818-294EA 7 USD
103818-294 103849-002 76770-610 76770-612 77087-512 77087-514 77087-516
5-Bromo-2-oxo-2,3-dihydropyridine-3-carbonitrile 98%
5-Bromo-2-oxo-2,3-dihydropyridine-3-carbonitrile

Precaución: for Research Use Only

Formula: C₆H₃BrN₂O
MW: 199.0048 g/mol
Boiling Pt: 292.1 °C (760 mm Hg)
Melting Pt: 219…221 °C
Flash Pt: 130.5 °C
Storage Temperature: Ambient
MDL Number: MFCD07363806
CAS Number: 405224-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR