5-Bromo-2-nitropyridine 98%

Supplier: Ambeed
Warning

A350557-25G A350557-5G A350557-1000G A350557-100G A350557-500G
103837-034EA 9 USD
103837-034 76840-138 76840-140 76989-628 76989-630 76989-632 76989-634
5-Bromo-2-nitropyridine 98%
5-Bromo-2-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₃BrN₂O₂
MW: 203 g/mol
Boiling Pt: 331.2 °C (760 mm Hg)
Flash Pt: 130.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00160411
CAS Number: 39856-50-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR