5-Bromo-2-nitrobenzaldehyde 97%

Supplier: Ambeed
Warning

A131972-100G A131972-10G A131972-250MG A131972-500G A131972-1G A131972-25G A131972-5G
103801-670EA 6 USD
103801-670 77009-994 77009-996 77009-998 77010-000 77314-206 77314-208 77314-210
5-Bromo-2-nitrobenzaldehyde 97%
5-Bromo-2-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrNO₃
MW: 230.0200 g/mol
Boiling Pt: 330.5 °C (760 mm Hg)
Flash Pt: 153.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00456508
CAS Number: 20357-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR