5-Bromo-2-nitroaniline 98%

Supplier: Ambeed
Warning

A556178-1G A556178-25G A556178-5G A556178-100G A556178-10G A556178-500G
103828-718EA 5 USD
103828-718 103839-854 76843-950 76843-952 76843-954 76980-250 76980-252 76980-254
5-Bromo-2-nitroaniline 98%
5-Bromo-2-nitroaniline

Precaución: For research use only

Formula: C₆H₅BrN₂O₂
MW: 217.0200 g/mol
Boiling Pt: 331.9 °C (760 mm Hg)
Flash Pt: 154.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09056836
CAS Number: 5228-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR