5-Bromo-2-naphthoic acid 95%

Supplier: Ambeed
Warning

A167398-10G A167398-250MG A167398-1G A167398-25G A167398-5G
103806-178EA 19 USD
103806-178 103844-440 76768-942 76768-944 77300-894 77300-896 77300-898
5-Bromo-2-naphthoic acid 95%
5-Bromo-2-naphthoic acid

Precaución: for Research Use Only

Formula: C₁₁H₇BrO₂
MW: 251.0800 g/mol
Boiling Pt: 387.3 °C (760 mm Hg)
Flash Pt: 188 °C
Storage Temperature: Ambient
MDL Number: MFCD08236734
CAS Number: 1013-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR