5-Bromo-2-methylpyrimidine 98%

Supplier: Ambeed
Warning

A459858-1G A459858-25G A459858-5G A459858-10G A459858-250MG
103827-908EA 6 USD
103827-908 103838-984 76842-858 76842-860 76842-862 77168-936 77168-938
5-Bromo-2-methylpyrimidine 98%
5-Bromo-2-methylpyrimidine

Precaución: for Research Use Only

Formula: C₅H₅BrN₂
MW: 173.01 g/mol
Boiling Pt: 195.3 °C (760 mm Hg)
Flash Pt: 71.885 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07375143
CAS Number: 7752-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR