5-Bromo-4-hydroxy-2-methylpyrimidine 96%

Supplier: Ambeed
Warning

A326832-100MG A326832-10G A326832-250MG A326832-25G A326832-1G A326832-5G
103825-450EA 54 USD
103825-450 103836-402 76773-008 76773-010 76773-012 76773-014 76839-372 76839-374
5-Bromo-4-hydroxy-2-methylpyrimidine 96%
5-Bromo-4-hydroxy-2-methylpyrimidine

Precaución: for Research Use Only

Formula: C₅H₅BrN₂O
Storage Temperature: Refrigerator
MDL Number: MFCD08460358
CAS Number: 1676-57-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR