5-Bromo-2-methylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-picoline, 3-Bromo-6-picoline

A259503-25G A259503-100G A259503-10G A259503-5G
103852-326EA 5 USD
103852-326 77091-896 77276-522 77276-524 77304-768
5-Bromo-2-methylpyridine 98%
5-Bromo-2-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 182.7 °C (760 mm Hg)
Melting Pt: 32…36 °C
Flash Pt: 64.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00661170
CAS Number: 3430-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR