5-Bromo-3-hydroxy-2-picoline 95%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-methylpyridin-3-ol

A107140-250MG A107140-25G A107140-1G A107140-5G A107140-100MG
103797-134EA 46 USD
103797-134 103822-894 76809-708 76809-710 77283-548 77283-550 77880-866
5-Bromo-3-hydroxy-2-picoline 95%
5-Bromo-3-hydroxy-2-picoline

Precaución: for Research Use Only

Formula: C₆H₆BrNO
MW: 188.0200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12024425
CAS Number: 91420-25-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR