5-Bromo-2-methylpyridin-3-amine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-5-bromo-2-methylpyridine

A180667-100G A180667-10G A180667-1G A180667-25G A180667-5G A180667-250MG
103807-548EA 6 USD
103807-548 103845-908 76769-542 76769-544 77083-406 77083-408 77083-410 77625-856
5-Bromo-2-methylpyridin-3-amine 98%
5-Bromo-2-methylpyridin-3-amine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂
MW: 187.04 g/mol
Flash Pt: 125.243 °C
Storage Temperature: Ambient
MDL Number: MFCD09031418
CAS Number: 914358-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR