5-Bromo-o-cresol 95%

Supplier: Ambeed
Warning

A326041-100G A326041-10G A326041-500G A326041-5G
103836-366EA 6 USD
103836-366 76772-988 76772-990 76772-992 77306-330
5-Bromo-o-cresol 95%
5-Bromo-o-cresol

Precaución: for Research Use Only

Formula: C₇H₇BrO
Boiling Pt: 241.8 °C (760 mm Hg)
Melting Pt: 76…78 °C
Flash Pt: 100.1 °C
Storage Temperature: Ambient
MDL Number: MFCD08061906
CAS Number: 36138-76-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR