5-Bromo-2-methyloxazole-4-carboxylic acid 97%

Supplier: Ambeed
Warning

A127821-100MG A127821-250MG A127821-1G
103801-018EA 115 USD
103801-018 76782-136 76782-138 77313-284
5-Bromo-2-methyloxazole-4-carboxylic acid 97%
5-Bromo-2-methyloxazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₄BrNO₃
MW: 205.9942 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09999146
CAS Number: 1209573-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR