5-Bromo-2-methylnicotinic acid 97%

Supplier: Ambeed
Warning

A214448-100MG A214448-250MG A214448-1G A214448-25G A214448-5G A214448-100G
103818-404EA 25 USD
103818-404 103849-110 76873-774 76873-776 77303-348 77303-350 77303-352 77612-108
5-Bromo-2-methylnicotinic acid 97%
5-Bromo-2-methylnicotinic acid

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
Melting Pt: 226…230 °C
Storage Temperature: Ambient
MDL Number: MFCD11557230
CAS Number: 351003-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR