5-Bromo-2-methylisoindolin-1-one 98%

Supplier: Ambeed
Warning

A215625-100MG A215625-10G A215625-250MG A215625-5G
103818-502EA 35 USD
103818-502 103849-210 77030-656 77030-658 77030-660 77303-414
5-Bromo-2-methylisoindolin-1-one 98%
5-Bromo-2-methylisoindolin-1-one

Precaución: for Research Use Only

Formula: C₉H₈BrNO
MW: 226.07 g/mol
Boiling Pt: 356.2 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD12755780
CAS Number: 868066-91-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR