5-Bromo-2-methylbenzaldehyde 97%

Supplier: Ambeed
Warning

A136538-250MG A136538-1G A136538-25G A136538-5G
103802-314EA 12 USD
103802-314 77109-572 77298-852 77315-222 77315-224
5-Bromo-2-methylbenzaldehyde 97%
5-Bromo-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇BrO
Flash Pt: 93.207 °C
Storage Temperature: Ambient
MDL Number: MFCD05662390
CAS Number: 90050-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR