5-Bromo-2-methyl-7-azaindole 97%

Supplier: Ambeed
Warning

A186669-100MG A186669-250MG A186669-1G A186669-5G
103815-954EA 48 USD
103815-954 103846-540 76974-668 76974-670 77084-214 77084-216
5-Bromo-2-methyl-7-azaindole 97%
5-Bromo-2-methyl-7-azaindole

Precaución: for Research Use Only

Formula: C₈H₇BrN₂
MW: 211.0600 g/mol
Melting Pt: 169…179 °C
Storage Temperature: Ambient
MDL Number: MFCD15529124
CAS Number: 1111638-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR