5-Bromo-3-nitro-2-picoline 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Bromo-2-methyl-3-nitropyridine
A187640-100G
A187640-10G
A187640-500G
A187640-25G
A187640-1G
A187640-5G
103816-028EA
5
USD
103816-028
103846-618
76784-808
76784-810
76784-812
77084-320
77302-090
77302-092
5-Bromo-3-nitro-2-picoline 97%
5-Bromo-3-nitro-2-picoline
Precaución: for Research Use Only
Formula:
C₆H₅BrN₂O₂ MW: 217.02 Boiling Pt: 252.6 °C (760 mm Hg) Flash Pt: 107 °C Storage Temperature: Ambient |
MDL Number:
MFCD09031419 CAS Number: 911434-05-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...